Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000625044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000625044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
ENST00000625044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000609684.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000625044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000625044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
ENST00000625044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013244.hg.1 |
NM_020957.3 |
Refseq |