Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002518.hg.1 |
TC09002930.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002518.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002518.hg.1 |
ENST00000377798.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002518.hg.1 |
ENST00000421959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002518.hg.1 |
ENST00000442009.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002518.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012124.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
ENST00000377792.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
ENST00000377798.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
ENST00000442009.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
NM_001282766.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
NM_022490.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183893.hg.1 |
TC0900012124.hg.1 |
Clariom D TC |