Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018906.2 | NP_061729.1 | 1 - 2 | 2389 | 71 | |
GenBank mRNA | AF152311.1 | - | 1 - 2 | 2389 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |