Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000243.2 | NP_000234.1 | 5 - 6 | 1627 | 63 | |
NM_001198536.1 | NP_001185465.1 | 4 - 5 | 994 | 63 | ||
XM_017023236.1 | XP_016878725.1 | 5 - 6 | 1765 | 63 | ||
GenBank mRNA | AF018080.1 | - | 5 - 6 | 1628 | 63 | |
AK298660.1 | - | 5 - 6 | 1621 | 63 | ||
BC101511.1 | - | 5 - 6 | 1612 | 8069017 | 63 | |
BC101537.1 | - | 5 - 6 | 1612 | 8069043 | 63 | |
FJ785717.1 | - | 2 - 3 | 508 | 63 | ||
FJ785722.1 | - | 4 - 5 | 954 | 63 | ||
Y14441.1 | - | 5 - 6 | 1616 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |