Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013110.hg.1 |
TC08001031.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013110.hg.1 |
ENST00000276420.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013110.hg.1 |
ENST00000544659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013110.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013112.hg.1 |
TC08001031.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013112.hg.1 |
ENST00000276420.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013112.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166676.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166676.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166676.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166676.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166676.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166676.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166676.hg.1 |
TC0800009819.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009819.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000276420.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000517422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000518197.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000522011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000523932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
ENST00000524001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
NM_003974.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800166674.hg.1 |
TC0800009819.hg.1 |
Clariom D TC |