Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000616802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
ENST00000616802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009148.hg.1 |
ENST00000345419.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009148.hg.1 |
ENST00000369878.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009148.hg.1 |
ENST00000433628.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009148.hg.1 |
ENST00000457502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009148.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009148.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009148.hg.1 |
TC10000769.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369875.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000475511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000616802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
ENST00000616802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008758.hg.1 |
NM_199077.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
ENST00000369878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
ENST00000433628.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
NM_017649.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
NM_199076.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159981.hg.1 |
TC1000008758.hg.1 |
Clariom D TC |