Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012926.hg.1 |
ENST00000281741.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012926.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012926.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012926.hg.1 |
TC14001140.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000554399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000555312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009184.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
ENST00000281741.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
ENST00000557374.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
NM_001160047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
NM_020784.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167774.hg.1 |
TC1400009184.hg.1 |
Clariom D TC |