Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011522359.1 | XP_011520661.1 | 2 - 3 | 928 | 76 | |
XM_011522360.1 | XP_011520662.1 | 2 - 3 | 928 | 76 | ||
GenBank mRNA | AK057178.1 | - | 4 - 5 | 610 | 76 | |
AY439221.1 | - | 4 - 5 | 404 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |