Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036775.hg.1 |
TC02002285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036775.hg.1 |
ENST00000322313.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036775.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203654.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203654.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203654.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203654.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203654.hg.1 |
TC0200014184.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000322313.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000393006.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000408998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000409658.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
ENST00000436787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006622.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_001006623.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014184.hg.1 |
NM_018383.4 |
Refseq |