Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000463746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000463746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000463746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000494384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000494384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000494384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000358204.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393481.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000393484.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000455989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000461440.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000463746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000463746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000463746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000485009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000492891.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000494384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000494384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000494384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
ENST00000496912.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_015641.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008868.hg.1 |
NM_152829.2 |
Refseq |