Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017016315.1 | XP_016871804.1 | 11 - 12 | 1762 | 106 | |
XM_017016316.1 | XP_016871805.1 | 11 - 12 | 2252 | 106 | ||
XM_017016318.1 | XP_016871807.1 | 10 - 11 | 1453 | 106 | ||
GenBank mRNA | AL713727.1 | - | 10 - 11 | 2008 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |