Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004309.hg.1 |
ENST00000288228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004309.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004309.hg.1 |
TC15000455.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000558900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000557914.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000558900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000559628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000560474.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
ENST00000561361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010749.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182606.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182606.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182606.hg.1 |
ENST00000288228.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182606.hg.1 |
NM_152450.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182606.hg.1 |
TC1500010749.hg.1 |
Clariom D TC |