Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023385.hg.1 |
TC04001686.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023385.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023385.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023384.hg.1 |
TC04001686.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023384.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023384.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012252.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184102.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184102.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184102.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184102.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184102.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184102.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184102.hg.1 |
TC0400012252.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
ENST00000307720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
ENST00000507213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
ENST00000512699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
NM_005038.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400184101.hg.1 |
TC0400012252.hg.1 |
Clariom D TC |