Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098514.2 | NP_001091984.2 | 2 - 3 | 587 | 89 | |
NM_152459.4 | NP_689672.4 | 2 - 3 | 587 | 89 | ||
XM_005255143.3 | XP_005255200.2 | 2 - 3 | 413 | 89 | ||
XM_017022972.1 | XP_016878461.1 | 2 - 3 | 413 | 89 | ||
XM_017022973.1 | XP_016878462.1 | 2 - 3 | 413 | 89 | ||
XM_017022974.1 | XP_016878463.1 | 2 - 3 | 413 | 89 | ||
GenBank mRNA | AK075242.1 | - | 2 - 3 | 413 | 89 | |
AK300708.1 | - | 2 - 3 | 361 | 89 | ||
AY358483.1 | - | 2 - 3 | 552 | 89 | ||
BC033681.1 | - | 2 - 3 | 482 | 5190553 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |