Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000390654.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000407622.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000484750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000484750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000484750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
ENST00000484750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013016.hg.1 |
NM_173465.3 |
Refseq |