Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005900.hg.1 |
TC02000393.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005900.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005900.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005899.hg.1 |
TC02000393.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005899.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005899.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005898.hg.1 |
TC02000393.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005898.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02005898.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154347.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154347.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154347.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154347.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154347.hg.1 |
TC0200007872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200154350.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154350.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154350.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154350.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154350.hg.1 |
TC0200007872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200154348.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154348.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154348.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154348.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154348.hg.1 |
TC0200007872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154349.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154349.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154349.hg.1 |
ENST00000272342.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154349.hg.1 |
NM_019002.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154349.hg.1 |
TC0200007872.hg.1 |
Clariom D TC |