Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174844.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174844.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174844.hg.1 |
TC0800011015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011015.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
ENST00000517981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
ENST00000523010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174840.hg.1 |
TC0800011015.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800174843.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174843.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174843.hg.1 |
ENST00000276616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174843.hg.1 |
NM_033126.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174843.hg.1 |
TC0800011015.hg.1 |
Clariom D TC |