Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052663.hg.1 |
TC01003366.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052663.hg.1 |
ENST00000292389.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052663.hg.1 |
ENST00000361516.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052663.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052663.hg.1 |
ENST00000392274.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052663.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368178.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
ENST00000469986.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016051.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221932.hg.1 |
TC0100016051.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
ENST00000361516.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
ENST00000368181.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
ENST00000462774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
NM_001159488.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
NM_030764.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
NR_125358.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221933.hg.1 |
TC0100016051.hg.1 |
Clariom D TC |