Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012069.hg.1 |
ENST00000330523.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012069.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012069.hg.1 |
TC20000887.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164616.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164616.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164616.hg.1 |
TC2000009239.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164616.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164616.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
ENST00000372643.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007495.hg.1 |
NM_080753.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
ENST00000335769.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009239.hg.1 |
NM_172131.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164615.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164615.hg.1 |
NM_172006.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164615.hg.1 |
TC2000009239.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164615.hg.1 |
ENST00000330523.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164615.hg.1 |
NM_172006.2 |
Refseq |