Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
TC01000723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
ENST00000371079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
ENST00000371080.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
ENST00000544776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
ENST00000545203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011491.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100160986.hg.1 |
TC0100008594.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000371080.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000482426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
ENST00000545203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_001083592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008594.hg.1 |
NM_005012.3 |
Refseq |