Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014825.2 | NP_055640.2 | 25 - 26 | 4410 | 80 | |
GenBank mRNA | AB011111.2 | - | 20 - 21 | 3644 | 80 | |
AF231919.1 | - | 25 - 26 | 4248 | 80 | ||
AF432264.1 | - | 3 - 4 | 431 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |