Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000299732.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000416617.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
ENST00000546692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011991.hg.1 |
NM_138451.2 |
Refseq |