Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130141.1 | NP_001123613.1 | 8 - 9 | 766 | 64 | |
GenBank mRNA | AY780793.1 | - | 8 - 9 | 689 | 64 | |
BC012061.1 | - | 8 - 9 | 755 | 4637826 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |