Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
TC06002022.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
ENST00000229470.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
ENST00000354650.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
ENST00000356013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
ENST00000368667.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
ENST00000368682.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
ENST00000544792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025284.hg.1 |
NM_002037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000354650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000368667.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000368682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000462598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000484067.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000487824.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000518295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000518630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000520518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000521062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000521361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000523238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000523570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000523574.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000524310.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
NM_002037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000354650.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000368667.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000368682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000462598.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000484067.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000487824.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000518295.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000518630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000520518.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000521062.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000521361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000523238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000523570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000523574.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
ENST00000524310.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
NM_002037.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188939.hg.1 |
TC0600012875.hg.1 |
Clariom D TC |