Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIVEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006734.3 | NP_006725.3 | 8 - 9 | 6363 | 73 | |
XM_017010805.1 | XP_016866294.1 | 8 - 9 | 6328 | 73 | ||
GenBank mRNA | AK226163.1 | - | 4 - 5 | 3841 | 73 | |
AK296531.1 | - | 4 - 5 | 611 | 73 | ||
CR936782.1 | - | 8 - 9 | 6363 | 73 | ||
DQ231041.1 | - | 4 - 5 | 5620 | 73 | ||
M61744.1 | - | 2 - 3 | 232 | 73 | ||
X65644.1 | - | 4 - 5 | 5797 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |