Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014308.hg.1 |
ENST00000313511.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014308.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014308.hg.1 |
TC16000994.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000577590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
ENST00000577590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NR_039872.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009883.hg.1 |
NR_039872.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000569217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000577590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
ENST00000577590.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NR_039872.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009883.hg.1 |
NR_039872.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
ENST00000313511.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
ENST00000561644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
ENST00000565012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
NM_003321.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600176057.hg.1 |
TC1600009883.hg.1 |
Clariom D TC |