Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
ENST00000313421.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
ENST00000344028.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
ENST00000346437.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
ENST00000352009.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
ENST00000452451.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
NM_001254750.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
NM_001254751.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
NM_006725.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
NR_045638.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006266.hg.1 |
TC11000523.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000313421.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000344931.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000352009.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000419282.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000433107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000452451.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000505761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NM_001254750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NM_001254751.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NM_006725.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NR_045638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000313421.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000344931.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000352009.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000419282.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000433107.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000452451.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
ENST00000505761.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NM_001254750.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NM_001254751.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NM_006725.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
NR_045638.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100155139.hg.1 |
TC1100007787.hg.1 |
Clariom D TC |