Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204514.1 | NP_001191443.1 | 15 - 16 | 2107 | 144 | |
NM_001204516.1 | NP_001191445.1 | 13 - 14 | 1811 | 144 | ||
NM_001204517.1 | NP_001191446.1 | 14 - 15 | 3052 | 144 | ||
NM_014970.3 | NP_055785.2 | 14 - 15 | 1943 | 144 | ||
XM_005244970.1 | XP_005245027.1 | 14 - 15 | 1946 | 144 | ||
XM_011509307.2 | XP_011507609.2 | 14 - 15 | 1946 | 144 | ||
GenBank mRNA | AK026471.1 | - | 14 - 15 | 1722 | 144 | |
AK122897.1 | - | 14 - 15 | 3054 | 144 | ||
AK297368.1 | - | 13 - 14 | 144 | |||
AK301912.1 | - | 7 - 8 | 910 | 144 | ||
AK303052.1 | - | 15 - 16 | 2064 | 144 | ||
AK315586.1 | - | 14 - 15 | 1857 | 144 | ||
BC028679.2 | - | 14 - 15 | 1900 | 4829354 | 144 | |
BC048012.1 | - | 13 - 14 | 1855 | 144 | ||
U59919.1 | - | 14 - 15 | 1860 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |