Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018766.hg.1 |
ENST00000289805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018766.hg.1 |
ENST00000335360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018766.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018766.hg.1 |
TC16001355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018765.hg.1 |
ENST00000289805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018765.hg.1 |
ENST00000335360.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018765.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018765.hg.1 |
TC16001355.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600187900.hg.1 |
TC1600011288.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600187904.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187904.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187904.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600187904.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187904.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600187904.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600187904.hg.1 |
TC1600011288.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000289805.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000335360.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
ENST00000569918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011288.hg.1 |
NM_152339.3 |
Refseq |