Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
TC05000830.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
ENST00000323668.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
ENST00000377797.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
ENST00000439160.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
ENST00000445265.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
ENST00000451292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
ENST00000504761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
ENST00000513346.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
NM_000356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
NM_001135243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
NM_001135244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
NM_001135245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011938.hg.1 |
NM_001195141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000323668.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000377797.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000427724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000439160.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000445265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000504761.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000506630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000513346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000514442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_000356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001135243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001135244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001135245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001195141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000323668.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000377797.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000427724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000439160.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000445265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000504761.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000506630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000513346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
ENST00000514442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_000356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001135243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001135244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001135245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
NM_001195141.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163494.hg.1 |
TC0500009088.hg.1 |
Clariom D TC |