Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016076.hg.1 |
TC0X001190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016076.hg.1 |
ENST00000262753.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016076.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016076.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169648.hg.1 |
TC0X00010207.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000262753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
ENST00000373145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_001307940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010207.hg.1 |
NM_024921.3 |
Refseq |