Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000468025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000468025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000468025.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000497509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
ENST00000497509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014921.hg.1 |
ENST00000262738.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014921.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014921.hg.1 |
TC22000889.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200165341.hg.1 |
TC2200009021.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165341.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165341.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165341.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165341.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165341.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165341.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000262738.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000454637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000468025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000468025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000468025.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000473624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000497509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
ENST00000497509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009021.hg.1 |
NM_014246.1 |
Refseq |