Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042544.1 | NP_001036009.1 | 23 - 24 | 3144 | 217 | |
NM_001042545.1 | NP_001036010.1 | 20 - 21 | 2960 | 217 | ||
NM_003573.2 | NP_003564.2 | 23 - 24 | 3033 | 217 | ||
XM_011527376.2 | XP_011525678.1 | 22 - 23 | 3287 | 217 | ||
XM_011527377.2 | XP_011525679.1 | 23 - 24 | 3192 | 217 | ||
XM_011527378.2 | XP_011525680.1 | 23 - 24 | 3192 | 217 | ||
XM_011527379.1 | XP_011525681.1 | 19 - 20 | 3095 | 217 | ||
XM_011527380.2 | XP_011525682.1 | 22 - 23 | 3066 | 217 | ||
XM_011527381.2 | XP_011525683.1 | 23 - 24 | 91 | |||
XM_011527382.2 | XP_011525684.1 | 22 - 23 | 2949 | 217 | ||
XM_011527383.2 | XP_011525685.1 | 23 - 24 | 3192 | 217 | ||
XM_011527384.2 | XP_011525686.1 | 23 - 24 | 91 | |||
XM_011527387.1 | XP_011525689.1 | 23 - 24 | 3292 | 217 | ||
XM_017027354.1 | XP_016882843.1 | 22 - 23 | 91 | |||
GenBank mRNA | AF051344.2 | - | 20 - 21 | 2960 | 217 | |
AF051345.2 | - | 23 - 24 | 3144 | 217 | ||
AK128191.1 | - | 9 - 10 | 1142 | 217 | ||
Y13622.1 | - | 22 - 23 | 3033 | 217 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |