Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000134.hg.1 |
TC08000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000134.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000134.hg.1 |
NM_018941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000136.hg.1 |
TC08000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000136.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000136.hg.1 |
NM_018941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000131.hg.1 |
TC08000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000131.hg.1 |
NM_018941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000132.hg.1 |
TC08000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000132.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000132.hg.1 |
NM_018941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000133.hg.1 |
TC08000008.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000133.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000133.hg.1 |
NM_018941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000519254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000519254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000519254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000523237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000523237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012256.hg.1 |
ENST00000523237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184494.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184494.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184494.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184494.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184494.hg.1 |
TC0800012256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800184491.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184491.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184491.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184491.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184491.hg.1 |
TC0800012256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800184493.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184493.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184493.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184493.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800184493.hg.1 |
TC0800012256.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000331222.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000519254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000519254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000519254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000520991.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000523237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000523237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012256.hg.1 |
ENST00000523237.1 |
Ensembl |