Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
TC06000185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
ENST00000356709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
ENST00000416795.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
ENST00000432533.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
ENST00000469230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
NM_001197237.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
NM_001197238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
NM_001197240.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001841.hg.1 |
NM_006995.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000352867.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000356709.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000416795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000432533.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000467485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000469230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000471116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000472507.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000482536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000482636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000483410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000493275.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000494184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197237.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197239.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_006995.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_181531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000352867.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000356709.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000416795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000432533.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000467485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000469230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000471116.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000472507.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000482536.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000482636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000483410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000493275.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
ENST00000494184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197237.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197239.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_001197240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_006995.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
NM_181531.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149483.hg.1 |
TC0600007302.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000356709.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000416795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000432533.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000469230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000482636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000493275.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000494184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_001197237.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_001197238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_001197240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_006995.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000356709.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000416795.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000432533.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000469230.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000482636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000493275.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
ENST00000494184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_001197237.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_001197238.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_001197240.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
NM_006995.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149484.hg.1 |
TC0600007302.hg.1 |
Clariom D TC |