Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000553758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000553758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006113.hg.1 |
ENST00000167106.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006113.hg.1 |
ENST00000556038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006113.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006113.hg.1 |
TC14000482.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400154882.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154882.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154882.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154882.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154882.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154882.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154882.hg.1 |
TC1400007732.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000167106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000553518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000553758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000553758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554237.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000554743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
ENST00000556038.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007732.hg.1 |
NM_014909.4 |
Refseq |