Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008134.hg.1 |
TC0X000618.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008134.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008134.hg.1 |
ENST00000443760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008134.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000432513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000432513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000432513.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157117.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157117.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157117.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00157117.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00157117.hg.1 |
TC0X00008388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000356892.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000432513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000432513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000432513.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
ENST00000476532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008388.hg.1 |
NM_018990.3 |
Refseq |