Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004450.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004450.hg.1 |
ENST00000280904.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004450.hg.1 |
ENST00000399347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004450.hg.1 |
ENST00000438199.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004450.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004450.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004450.hg.1 |
TC18000436.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800163958.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163958.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163958.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163958.hg.1 |
TC1800009268.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800163958.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800163958.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800163958.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
ENST00000251081.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_004949.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009268.hg.1 |
NM_024422.4 |
Refseq |