Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006290.hg.1 |
TC06004084.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006290.hg.1 |
ENST00000341376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006290.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006290.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006290.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014123.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
ENST00000341376.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
ENST00000353205.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
NM_002505.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
NM_021705.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198592.hg.1 |
TC0600014123.hg.1 |
Clariom D TC |