Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003406.hg.1 |
TC04000228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003406.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003406.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149821.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149821.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149821.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149821.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149821.hg.1 |
TC0400007282.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
ENST00000257408.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007282.hg.1 |
NM_175737.3 |
Refseq |