Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013593.hg.1 |
TC09001037.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013593.hg.1 |
ENST00000303992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013593.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013593.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013594.hg.1 |
TC09001037.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013594.hg.1 |
ENST00000303992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013594.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09013594.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167861.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167861.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167861.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167861.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167861.hg.1 |
TC0900009902.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009902.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167862.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167862.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167862.hg.1 |
ENST00000399775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900167862.hg.1 |
NM_198573.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900167862.hg.1 |
TC0900009902.hg.1 |
Clariom D TC |