Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000690.hg.1 |
TC0X000051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000690.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000690.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000692.hg.1 |
TC0X000051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000692.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000692.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000692.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145316.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145316.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145316.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145316.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145316.hg.1 |
TC0X00006625.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006625.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145318.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145318.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145318.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145318.hg.1 |
ENST00000380659.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145318.hg.1 |
ENST00000484204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145318.hg.1 |
NM_016562.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145318.hg.1 |
TC0X00006625.hg.1 |
Clariom D TC |