Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018027.hg.1 |
ENST00000159087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018027.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19018027.hg.1 |
TC19001273.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000600711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009963.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
NM_020959.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
TC1900009963.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
ENST00000159087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
ENST00000597643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
ENST00000630631.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900181787.hg.1 |
NM_020959.2 |
Refseq |