Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012433.3 | NP_036565.2 | 17 - 18 | 2586 | 67 | |
GenBank mRNA | AF054284.1 | - | 17 - 18 | 2492 | 67 | |
AK308122.1 | - | 17 - 18 | 2539 | 67 | ||
BE168209.1 | - | 2 - 3 | 67 | |||
BE168220.1 | - | 2 - 3 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |