Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027840.hg.1 |
TC02001730.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027840.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027840.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189150.hg.1 |
TC0200012220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200189151.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189151.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189151.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189151.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189151.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189151.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189151.hg.1 |
TC0200012220.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000490394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
ENST00000491596.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012220.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189152.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189152.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189152.hg.1 |
ENST00000474610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200189152.hg.1 |
NR_003143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200189152.hg.1 |
TC0200012220.hg.1 |
Clariom D TC |