Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000483902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000483902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000483902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037485.hg.1 |
TC02002376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037485.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037485.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200205015.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205015.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200205015.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205015.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200205015.hg.1 |
TC0200014404.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000264156.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000483902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000483902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000483902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
ENST00000492091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014404.hg.1 |
NM_005915.5 |
Refseq |