Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028802.hg.1 |
TC01001851.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028802.hg.1 |
ENST00000430092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028802.hg.1 |
ENST00000439375.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028802.hg.1 |
NM_001373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
ENST00000430092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
ENST00000439375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
ENST00000495456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
NM_001373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
ENST00000430092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
ENST00000439375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
ENST00000495456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
NM_001373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186720.hg.1 |
TC0100011755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000430092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000439375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000445597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000495456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
NM_001373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000430092.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000439375.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000445597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
ENST00000495456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
NM_001373.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100186719.hg.1 |
TC0100011755.hg.1 |
Clariom D TC |