Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004568.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004568.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004568.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004568.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004568.hg.1 |
TC21000402.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000382373.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000479548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000487113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000490358.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007968.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
TC2100007968.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153729.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
NM_019596.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
TC2100007968.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
ENST00000536776.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
ENST00000624800.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
NM_001162495.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
NM_001162496.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153728.hg.1 |
NM_019596.5 |
Refseq |