Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077526.2 | NP_001070994.1 | 17 - 18 | 1762 | 111 | |
XM_017007036.1 | XP_016862525.1 | 18 - 19 | 1931 | 111 | ||
XM_017007037.1 | XP_016862526.1 | 17 - 18 | 1857 | 111 | ||
XM_017007038.1 | XP_016862527.1 | 16 - 17 | 1660 | 111 | ||
XM_017007043.1 | XP_016862532.1 | 16 - 17 | 1531 | 111 | ||
XM_017007045.1 | XP_016862534.1 | 15 - 16 | 1567 | 111 | ||
GenBank mRNA | AF289603.1 | - | 17 - 18 | 1726 | 111 | |
BC050626.1 | - | 17 - 18 | 1685 | 4416416 | 111 | |
DQ630520.1 | - | 17 - 18 | 1611 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |