Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193285.2 | NP_001180214.1 | 5 - 6 | 597 | 73 | |
NM_001321061.1 | NP_001307990.1 | 4 - 5 | 469 | 73 | ||
GenBank mRNA | BC004218.2 | - | 4 - 5 | 409 | 3352203 | 73 |
BC028220.1 | - | 5 - 6 | 543 | 5217599 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |